Principaux documents
TEM-CF300-AU-ET
Sigma-Aldrich
Carbon Film Supported Gold Square Mesh
Extra Thick, size 300 mesh, box of 50
About This Item
Code UNSPSC :
41111761
Taille:
300 mesh
Conditionnement:
box of 50
Conditionnement
box of 50
Taille
300 mesh
Description générale
A 20-30 nm thick film of pure carbon supported standard squre mesh grids. Gold grid, 300 mesh.