V800181
Ethylene glycol
LR, ≥99%
Synonym(s):
1,2-Ethanediol
About This Item
Recommended Products
grade
LR
vapor density
2.1 (vs air)
vapor pressure
0.08 mmHg ( 20 °C)
product line
Vetec™
Assay
≥99%
form
liquid
autoignition temp.
752 °F
expl. lim.
15.3 %
dilution
(for analytical testing)
refractive index
n20/D 1.431 (lit.)
bp
195-198 °C
mp
−13 °C (lit.)
density
1.113 g/mL at 25 °C (lit.)
SMILES string
OCCO
InChI
1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2
InChI key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - STOT RE 2 Oral
Target Organs
Kidney
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
239.0 °F - open cup
Flash Point(C)
115 °C - open cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service