Skip to Content
Merck
All Photos(1)

Key Documents

G003907

GC Analysis of Fuel Oil #4 on Equity-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

suitable for GC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

gas chromatography (GC): suitable

test parameters

carrier gas: hydrogen, 45 cm/sec constant
column: Equity-1, 15 m x 0.10 mm I.D., 0.10 μm (28039-U)
detector: FID, 350 °C
inj. temp.: 250 °C
injection: 0.3 μL, 200:1 split, 0.02 min pre-injection dwell time
liner: 2 mm I.D., straight
oven: 80 °C, 50 °C/min to 325 °C
sample: No.4 Fuel Oil standard, 50 mg/mL in hexane (47535-U)

application(s)

petroleum

Legal Information

analytical column


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service