Skip to Content
Merck
All Photos(1)

Key Documents

SRP3121

Sigma-Aldrich

NAP-2 (CXCL7) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CTAP-III (precursor), CXCL7, Neutrophil Activating Protein-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

7.6 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PPBP(5473)

General description

NAP-2 is a CXC chemokine that can signal through the CXCR1 and CXCR2 receptors. It is produced in leukocytes by enzymatic processing of a precursor called platelet basic protein (PBP). NAP-2 chemoattracts and activates neutrophils. Recombinant human NAP-2 protein is a 7.6 kDa protein containing 70 amino acid residues including the four highly conserved cysteine residues present in CXC chemokines, and also including the "ELR" motif common to CXC chemokines that bind to CXCR1 and CXCR2.

Biochem/physiol Actions

NAP-2 is a CXC chemokine that can signal through the CXCR1 and CXCR2 receptors. Recombinant human NAP-2 protein is a 7.6 kDa protein containing 70 amino acid residues including the four highly conserved cysteine residues present in CXC chemokines, and also including the "ELR" motif common to CXC chemokines that bind to CXCR1 and CXCR2.

Sequence

AELRCMCIKT TSGIHPKNIQ SLEVIGKGTH CNQVEVIATL KDGRKICLDP DAPRIKKIVQ KKLAGDESAD

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brandt, E., et al.
Cytokine Reference, 1069-1094 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service