Skip to Content
Merck
All Photos(2)

Key Documents

SML3905

Sigma-Aldrich

FerroLOXIN-2

≥98% (HPLC)

Synonym(s):

N-(3-Bromo-4-(trifluoromethyl)benzyl)-1-(3-fluorophenyl)-4-phenyl-1H-imidazol-2-amine trifluoroacetate, N-[[3-Bromo-4-(trifluoromethyl)phenyl]methyl]-1-(3-fluorophenyl)-4-phenyl-1H-imidazol-2-amine trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H16BrF4N3 ·CF3CO2H
CAS Number:
Molecular Weight:
604.31
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

15LOX/PEBP1 catalytic complex inhibitor that prevents cell death by ferroptosis, but not necroptosis, pyroptosis, or apoptosis.
FerroLOXIN-2 is a ferroptosis inhibitor that selectively targets the 15LOX/PEBP1 catalytic complex without affecting the biosynthesis of pro-/anti-inflammatory lipid mediators. FerroLOXIN-2 prevents 0.5 μM GPX4 inhibitor RSL3-induced ferroptosis in HBE cell cultures (IC50 = 97 nM post 20h RSL3 treatment) and rescues mice from total body irradiation (TBI)-induced death in vivo (Cpd administered via 25 mg/kg ip. 20h post 9.25Gy irradiation; survival on day 30 = 80% vs. 0% on day 19 of untreated control). FerroLOXIN-2 does not exert its efficacy via radical scavenging or iron-chelating activity, nor does it prevent cell death by necroptosis, pyroptosis, or apoptosis.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service