Skip to Content
Merck
All Photos(1)

Key Documents

SAB4702025

Monoclonal Anti-EGFR PE produced in mouse

new

Synonym(s):

EGF-R, ERBB1, Her1, NISBD2, PIG61

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

phycoerythrin (R-PE) conjugate

Quality Level

antibody product type

primary antibodies

clone

EGFR1, monoclonal

form

buffered aqueous solution

species reactivity

horse, ( Human)

technique(s)

flow cytometry: suitable

isotype

IgG2bκ

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... EGFR(1956)

General description

The oncoprotein EGFR (epidermal growth factor receptor), also known as HER1 / ErbB1, is a member of ErbB family of cell surface receptor tyrosine kinases. This 170 kDa transmembrane glycoprotein is often associated with cancerogenesis, although its activation state is controlled at various levels including trafficking and degradation steps. Binding of receptor-specific ligands to the EGFR ectodomain results in formation of homodimeric or heterodimeric kinase-active complexes into which HER2 / ErbB2 is recruited as a preferred partner. Subsequent signaling cascades such as RAS/MAPK and PI3K/AKT pathways lead to cell proliferation and survival responses.

Specificity

The mouse monoclonal antibody EGFR1 reacts with extracellular domain of human protein kinase EGFR (ErbB1 / HER1); epitope within amino acids 6-273.

Immunogen

Human epidermoid carcinoma line A431

Physical form

Stabilizing phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with R-phycoerythrin (PE) under optimum conditions. Unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service