Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300512

Sigma-Aldrich

Anti-Mouse Dnmt2 (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-DNMT2, Anti-M.MmuIIP, Anti-Putative DNA cytosine methyltransferase DNMT2, DNA MTase homolog MmuIIP, DNA methyltransferase homolog MmuIIP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Trdmt1(13434)

General description

mDnmt2 does not appear to be active as a DNA methyltransferase; however, its strong binding to DNA suggests that it may mark specific sequences in the genome by binding to DNA through the specific target-recognizing motif. mDnmt2 is strongly expressed in thymus, testis, and at much lower levels in spleen, lung, brain, heart, kidney, liver, skeletal muscle and embryonic stem cells. This protein belongs to the 5-cystosine methyltransferase family.

Immunogen

DNMT2 (351-386)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of mouse Dnmt2.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service