Skip to Content
Merck
All Photos(1)

Key Documents

I9898

Sigma-Aldrich

Anti-Interleukin-5 antibody produced in goat

IgG fraction of antiserum, lyophilized powder

Synonym(s):

Anti-IL-5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (i)
Neutral
WB
clone:
polyclonal
species reactivity:
human
citations:
3
technique(s):
indirect ELISA: 0.5-1.0 μg/test
neutralization: suitable
western blot: 1-2 μg/mL

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

indirect ELISA: 0.5-1.0 μg/test
neutralization: suitable
western blot: 1-2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL5(3567)

General description

The antibody neutralizes the biological activity of recombinant human IL-5.

Immunogen

recombinant human IL expressed in Sf 21 insect cell line.

Biochem/physiol Actions

Interleukin-5 is a T-cell derived cytokine that induces the differentiation of B cells into antibody producing plasma cells. It is a critical differentiation factor for eosinophils and regulates the expression of genes required for cell proliferation, survival and maturation. The cellular functions of IL-5 are mediated by JAK-STAT, Btk and Ras/Raf-ERK signaling pathways. IL-5 is a good molecular target for the treatment of eosinophilic diseases.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Preparation Note

Purified using protein G.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taku Kouro et al.
International immunology, 21(12), 1303-1309 (2009-10-13)
IL-5 was originally defined as a T-cell-derived cytokine that triggers activated B cells for terminal differentiation into antibody-secreting plasma cells, at least in mice. Concurrently, IL-5 was recognized as the major maturation and differentiation factor for eosinophils in mice and
Kiyoshi Takatsu
Proceedings of the Japan Academy. Series B, Physical and biological sciences, 87(8), 463-485 (2011-10-12)
While interleukin-5 (IL-5) is initially identified by its ability to support the growth and terminal differentiation of mouse B cells in vitro into antibody-secreting cells, recombinant IL-5 exerts pleiotropic activities on various target cells including B cells, eosinophils, and basophils.
Jonathan Corren
Discovery medicine, 13(71), 305-312 (2012-05-01)
Elevated numbers of blood and tissue eosinophils are present in allergic diseases and experimental evidence suggests that eosinophils play an important pathogenic role in these conditions. Regulation of eosinophil maturation, recruitment, and survival is under the control of a small
Patricia C Fulkerson et al.
Journal of immunology (Baltimore, Md. : 1950), 193(8), 4043-4052 (2014-09-19)
Eosinophils originate in the bone marrow from an eosinophil lineage-committed, IL-5Rα-positive, hematopoietic progenitor (eosinophil progenitor). Indeed, IL-5 is recognized as a critical regulator of eosinophilia and has effects on eosinophil progenitors, eosinophil precursors, and mature eosinophils. However, substantial levels of
Qiong Long et al.
Human vaccines & immunotherapeutics, 10(8), 2303-2311 (2014-11-27)
We sought to develop an IL-33 vaccine and evaluate its efficacy in a mouse model of asthma. The full-length molecules of putative mature IL-33 were inserted into the immunodominant epitope region of hepatitis B core antigen using gene recombination techniques.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service