Skip to Content
Merck
All Photos(2)

Key Documents

HPA023399

Sigma-Aldrich

Anti-ARFGEF1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Brefeldin A-inhibited GEP 1, Anti-Brefeldin A-inhibited guanine nucleotide-exchange protein 1, Anti-p200 ARF guanine nucleotide exchange factor, Anti-p200 ARF-GEP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

VLYDGENHDCEEKPQDIVQNIVEEMVNIVVGDMGEGTTINASADGNIGTIEDGSDSENIQANGIPGTPISVAYTPSLPDDRLSVSSNDTQESGNSSGPSPGAKF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARFGEF1(10565)

Related Categories

General description

The gene ARFGEF1 (ADP ribosylation factor guanine nucleotide exchange factor 1), also referred to as BIG1 (brefeldin A-inhibited guanine nucleotide-exchange protein), is a guanine nucleotide exchange factor that acts on trans-Golgi. The gene is mapped to human chromosome 8.

Immunogen

Brefeldin A-inhibited guanine nucleotide-exchange protein 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-ARFGEF1 antibody produced in rabbit has been used for immunofluorescence. All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene ARFGEF1 (ADP ribosylation factor guanine nucleotide exchange factor 1) encodes an ADP-ribosylation factor that catalyzes the replacement of bound GDP with GTP via the activation of class I ADP-ribosylation factors (ARF1-3). This process is necessary for the regulation of protein transport in eukaryotic cells. It also functions as a scaffolding protein and interacts with various proteins in other compartments of the cell. Knock-down of BIG1 and BIG2, a closely related guanine-nucleotide exchange factor, results in disruption of localization of certain proteins associated with the trans-Golgi network (TGN) and recycling endosomes. The retrograde transport of furin from late endosomes to the TGN is also inhibited in the absences of these two proteins.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76183

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dylan Mordaunt et al.
Pediatric neurology, 52(2), 230-234 (2015-02-20)
Cerebellar vermis hypoplasia has been associated with a large number of chromosomal abnormalities and metabolic disorders, with few candidate genes clearly linked to isolated cerebellar vermis hypoplasia. We describe on a 12-year-old boy with inferior vermian hypoplasia associated with a
Chantal Christis et al.
The Journal of cell biology, 196(3), 327-335 (2012-02-01)
The small G protein Arf1 regulates Golgi traffic and is activated by two related types of guanine nucleotide exchange factor (GEF). GBF1 acts at the cis-Golgi, whereas BIG1 and its close paralog BIG2 act at the trans-Golgi. Peripheral membrane proteins
Ray Ishizaki et al.
Molecular biology of the cell, 19(6), 2650-2660 (2008-04-18)
BIG2 and BIG1 are closely related guanine-nucleotide exchange factors (GEFs) for ADP-ribosylation factors (ARFs) and are involved in the regulation of membrane traffic through activating ARFs and recruiting coat protein complexes, such as the COPI complex and the AP-1 clathrin
Ju Hee Kim et al.
BMB reports, 44(8), 523-528 (2011-08-30)
To identify novel genes that are regulated by promoter methylation, a combinational approach involving in silico mining followed by molecular assay was performed. From the expression microarray data registered in the European bioinformatics institute (EBI), genes showing downregulation in breast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service