Skip to Content
Merck
All Photos(5)

Key Documents

HPA005550

Sigma-Aldrich

Anti-PROC antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Anticoagulant protein C antibody produced in rabbit, Anti-Autoprothrombin IIA antibody produced in rabbit, Anti-Blood coagulation factor XIV antibody produced in rabbit, Anti-Vitamin K-dependent protein C precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
human
citations:
8
technique(s):
immunohistochemistry: 1:50- 1:200

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

AHCMDESKKLLVRLGEYDLRRWEKWELDLDIKEVFVHPNYSKSTTDNDIALLHLAQPATLSQTIVPICLPDSGLAERELNQAGQETLVTGWGYHSSREKEAKRNRTFVLNFIKIPVVPHNECSEVMSNMVSENMLCAGI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PROC(5624)

General description

PROC (protein C) is an anticoagulant and a serine protease. This gene is localized to chromosome 2q13-q14. This protein belongs to a family of proteins which are vitamin-K dependent. PROC is a precursor to activated protein C (APC), which is a glycoprotein, and is produced by thrombin cleavage of an activation signal. It is expressed by endothelial cells, keratinocytes and certain hematopoietic cells. It is also expressed in liver.

Immunogen

Vitamin K-dependent protein C precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Protein C (PROC) forms the anti-coagulation pathway, along with the endothelial cell protein C receptor (EPCR), thrombin, thrombomodulin, and protein S. This pathway is responsible for controlling thrombosis and inflammatory response. It is also thought to decrease endothelial cell death during ischemia and inflammation. It inhibits factors Va and VIIIa, and activates fibrinolysis, to prevent coagulation. There are two types of hereditary PROC deficiency- type I, which has both decreased antigen concentration and enzyme activity, and type II, which has only decreased enzyme activity. These deficiencies result in thrombotic diseases. Compound heterozygous or homozygous deficiencies in PROC gene leads to purpura fulminans and severe form of thromboembolic disease, beginning from the perinatal period. This protein determines the outcome of multiple inflammatory diseases, such as encephalitis and sepsis. In diabetic mice, this protein protects against diabetic nephropathy. It also has potential as a marker for the early diagnosis of diabetic nephropathy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83061

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hartmut Weiler
Thrombosis research, 133 Suppl 1, S32-S34 (2014-04-25)
The inflammation-induced activation of the protein C pathway provides negative feedback inhibition of coagulation and exerts coagulation-independent anti-inflammatory and cytoprotective effects. The balance between these activities of aPC modulates the outcome of diverse inflammatory diseases such as encephalitis, diabetes, and
Weiping Li et al.
Clinical laboratory, 60(2), 261-266 (2014-03-26)
Activated protein C (APC) plays a vital renoprotective role against diabetic nephropathy in STZ-induced diabetic mice by inhibiting endothelial cell and podocyte apoptosis. This study aimed to examine the relationship between the degree of albuminuria and APC levels in patients
Christopher J Jackson et al.
The international journal of biochemistry & cell biology, 40(12), 2692-2697 (2008-02-06)
Activated protein C (APC) is a glycoprotein derived from its precursor, protein C and formed by the cleavage of an activation peptide by thrombin bound to thrombomodulin. Originally thought to be synthesized exclusively by the liver, recent reports have shown
G Romeo et al.
Proceedings of the National Academy of Sciences of the United States of America, 84(9), 2829-2832 (1987-05-01)
The structure of the gene for protein C, an anticoagulant serine protease, was analyzed in 29 unrelated patients with hereditary thrombophilia and protein C deficiency. Gene deletion(s) or gross rearrangement(s) was not demonstrable by Southern blot hybridization to cDNA probes.
P Patracchini et al.
Human genetics, 81(2), 191-192 (1989-01-01)
The localization of human protein C gene on chromosome 2 was investigated by in situ hybridization using a partial cDNA for protein C. Silver-grain analysis indicates that the protein C gene is located on 2q13-q14.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service