HP 6054
Reactivity: Anti-λ-Light chain, human, 93092704
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
Mouse x Mouse Hybridoma
ImmunoDon
BALB/c mouse spleen
Myeloma
Sp2/0 Ag-14
Reactivity
Anti-λ-Light chain, human
technique(s)
cell culture | mammalian: suitable
isotype
IgG2a
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse Sp2/0 Ag-14 x Mouse BALB/c spleen
Cell Line Description
The antibody is directed against human kappa light chains.
Immunogen
Purified human immunoglobulin myeloma protein covalently coupled to polyaminostyrene beads
Application
Classification of human imunoglobulin
Culture Medium
DMEM + 2mM glutamine + 10mM HEPES + 0.05mM 2-mercaptoethanol (2ME) + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 100,000 - 1,000,000 cells/ml; 5% CO2; 37°C.
Other Notes
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service