Recommended Products
biological source
Mouse x Mouse Hybridoma
ImmunoDon
BALB/c mouse spleen
Myeloma
Sp2/0
Reactivity
Anti-IgG, grey seal
technique(s)
cell culture | mammalian: suitable
isotype
IgG1
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Not specified
Cell Line Description
The antibody is directed against seal IgG.
Immunogen
Grey seal immunoglobulin
Application
Quantification of seal IgG
Culture Medium
RPMI 1640 + 2 mM glutamine + 10% Fetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37 °C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service