Skip to Content
Merck
All Photos(1)

Key Documents

AXYPCR384M2CS

PCR microplate for ABI 3730, 3100 and MJ Thermocyclers

384 well, sterile

Synonym(s):

384 multiwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800

material

clear plate
clear polypropylene wells

sterility

sterile

packaging

case of 50 ea

manufacturer/tradename

Axygen Inc
Corning

technique(s)

PCR: suitable

size

384 wells

working volume

50 mL

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service