Skip to Content
Merck
All Photos(1)

Key Documents

357413

Sigma-Aldrich

Sulfuric acid, fuming

reagent grade, 30% free SO3 basis

Synonym(s):

Oleum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4 · (SO3)x
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 25 °C)

form

liquid (viscous, may be cloudy)

concentration

30% (free SO3)

density

1.925 g/mL at 25 °C (lit.)

SMILES string

O=S(=O)=O.OS(O)(=O)=O

InChI

1S/H2O4S.O3S/c1-5(2,3)4;1-4(2)3/h(H2,1,2,3,4);

InChI key

HIFJUMGIHIZEPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

may contains particles

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service