Skip to Content
Merck
All Photos(1)

Key Documents

34994

Sigma-Aldrich

Hexane, mixture of isomers

≥85%, suitable for HPLC: suitable

Synonym(s):

Hexanes, Isohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

Product Name

Hexane, mixture of isomers, suitable for HPLC, ≥85%

vapor density

3 (vs air)

Assay

≥85%

autoignition temp.

437 °F

expl. lim.

7.5 %

technique(s)

HPLC: suitable

impurities

≤0.0005% non-volatile matter
≤0.001% free acid (as CH3COOH)
≤0.01% water (Karl Fischer)
≤0.5 ppb fluorescence (quinine) at 254 nm
≤0.5 ppb fluorescence (quinine) at 365 nm
≤4.9% n-hexane (GC)

evapn. residue

≤0.0005%

transmittance

200 nm, ≥20%
220 nm, ≥80%
245 nm, ≥98%

refractive index

n20/D 1.379 (lit.)

bp

68-70 °C (lit.)

mp

−95 °C (lit.)

density

0.672 g/mL at 25 °C (lit.)

SMILES string

C(CCC)(C)C

InChI

1S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3

InChI key

AFABGHUZZDYHJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

19.4 °F

Flash Point(C)

-7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extraction and Identification of T Cell Stimulatory Self-lipid Antigens.
Lepore M, et al.
Bio-protocol, 5(11) (2015)
Eagleson M.
Concise Encyclopedia Chemistry, 492-492 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service