Skip to Content
Merck
All Photos(1)

Key Documents

W381812

Sigma-Aldrich

L-Alanine

natural, FG

Synonym(s):

(S)-2-Aminopropionic acid, L-α-Aminopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7NO2
CAS Number:
Molecular Weight:
89.09
FEMA Number:
3818
Beilstein:
1720248
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Flavis number:
17.002
Organoleptic:
sweet
grade:
FG
Halal
Kosher
natural
biological source:
corn
food allergen:
no known allergens

biological source

corn

Quality Level

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.320

application(s)

flavors and fragrances

food allergen

no known allergens

Organoleptic

sweet

SMILES string

C[C@H](N)C(O)=O

InChI

1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m0/s1

InChI key

QNAYBMKLOCPYGJ-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Patrycja Puchalska et al.
Cell metabolism, 29(2), 383-398 (2018-11-20)
Metabolic plasticity has been linked to polarized macrophage function, but mechanisms connecting specific fuels to tissue macrophage function remain unresolved. Here we apply a stable isotope tracing, mass spectrometry-based untargeted metabolomics approach to reveal the metabolome penetrated by hepatocyte-derived glucose and
Markus Mund et al.
Cell, 174(4), 884-896 (2018-07-31)
Clathrin-mediated endocytosis is an essential cellular function in all eukaryotes that is driven by a self-assembled macromolecular machine of over 50 different proteins in tens to hundreds of copies. How these proteins are organized to produce endocytic vesicles with high precision

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service