EZFKIT001SZ
EZ-Fluo™ System
Contains EZ-Fluo™ reader, EZ-Fluo™ camera, power cord for Switzerland
Synonym(s):
Detection instrument, EZ-Fluo™ System Kit (SZ), For rapid detection of microbial contamination in filterable samples, suitable for bioburden testing
About This Item
AC/DC input
100 V / 240 V, 50 - 60 Hz
Quality Level
manufacturer/tradename
EZ-Fluo™
parameter
4.4 kg at 15 - 40 °C
H × W × L
12.5 cm × 14.2 cm × 24.9 cm
application(s)
environmental
food and beverages
pharmaceutical
compatibility
EZ-Fluo™ Rapid Microbial Detection System
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Rapid technology capable of providing results in 1/3 of the time required using traditional techniques
- Flexible approach based on standard membrane filtration and reduced incubation on media plate
- Non-destructive method, compatible with any ID technique
- Compact hardware, fits easily on any laboratory bench
- Cost effective solution for both routine and investigatory use
Components
- EZ-Fluo™ System standard kit
- 1 reader
- 1 camera
- 1 software
Legal Information
related product
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service