Skip to Content
Merck
All Photos(1)

Key Documents

6.17945

Supelco

Zinc sulfate heptahydrate

GR

Synonym(s):

Zinc sulfate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSO4 · 7H2O
CAS Number:
Molecular Weight:
287.56
MDL number:
UNSPSC Code:
12352302
grade:
for analytical purposes

grade

for analytical purposes

SMILES string

O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.7H2O.Zn/c1-5(2,3)4;;;;;;;;/h(H2,1,2,3,4);7*1H2;/q;;;;;;;;+2/p-2

InChI key

RZLVQBNCHSJZPX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service