Skip to Content
Merck
All Photos(1)

Key Documents

551590

Sigma-Aldrich

Quercetagetin

A cell-permeable flavanol compound that acts a potent, ATP-competitive, and reversible inhibitor of PIM1 kinase (IC₅₀ = 340 nM).

Synonym(s):

Quercetagetin, 3,3ʹ,4ʹ,5,6,7-Hexahydroxyflavone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O8
CAS Number:
Molecular Weight:
318.24
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow to yellow-green

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C15H10O8/c16-6-2-1-5(3-7(6)17)15-14(22)13(21)10-9(23-15)4-8(18)11(19)12(10)20/h1-4,16-20,22H

InChI key

ZVOLCUVKHLEPEV-UHFFFAOYSA-N

General description

A cell-permeable flavanol compound that acts a potent, ATP-competitive, and reversible inhibitor of PIM1 kinase (IC50 = 340 nM). Exhibits ≥7-fold selectivity over 8 other kinases tested, including PIM2 (IC50 = 3.45 µM), and little activity toward c-Abl even at concentrations as high as 200 µM. Inhibits cellular BAD phosphorylation (IC50 = 5.5 µM) and induces growth arrest (ED50 = 3.8 µM) in PIM1-expressing RWPE2 cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PIM1 kinase
Product competes with ATP.
Reversible: yes
Target IC50: 340 nM against PIM1 kinase

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic & Carcinogenic / Teratogenic (G)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Holder, S., et al. 2007. Mol. Cancer Ther.6, 163.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service