325001
Epothilone B, Synthetic
A structural analog of Epothilone A with similar biological properties.
Synonym(s):
Epothilone B, Synthetic, EpoB
About This Item
Recommended Products
Quality Level
Assay
≥99% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white to off-white
solubility
DMSO: 10 mg/mL
shipped in
ambient
storage temp.
−20°C
InChI
1S/C27H41NO6S/c1-15-9-8-10-27(7)22(34-27)12-20(16(2)11-19-14-35-18(4)28-19)33-23(30)13-21(29)26(5,6)25(32)17(3)24(15)31/h11,14-15,17,20-22,24,29,31H,8-10,12-13H2,1-7H3/b16-11+/t15-,17+,20-,21-,22-,24-,27+/m0/s1
InChI key
QXRSDHAAWVKZLJ-PVYNADRNSA-N
General description
Biochem/physiol Actions
P-glycoprotein-expressing multidrug resistant (MDR) cells
Warning
Reconstitution
Other Notes
Chou, T.-C., et al. 1998. Proc. Natl. Acad. Sci. USA 95, 9642.
Kowalski, R.J., et al. 1997. J. Biol. Chem. 272, 2534.
Muhlradt, P.F., and Sasse, F. 1997. Cancer Res. 57, 3344.
Nicolaou, K.C., et al. 1997. Nature 387, 268.
Bollag, D.M., et al. 1995. Cancer Res. 55, 2325.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service