Skip to Content
Merck
All Photos(1)

Key Documents

252721

Millipore

n-Decanoylsucrose

A non-ionic detergent useful for solubilization and isolation of pigment proteins of brown algae. Can be easily removed by dialysis.

Synonym(s):

n-Decanoylsucrose, α-D-Glucopyranoside, β-D-Fructofuranosyl Monodecanoate, Sucrose Monocaprate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H40O12
CAS Number:
Molecular Weight:
496.55
MDL number:
UNSPSC Code:
12161902

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white to off-white

CMC

2.5 mM

solubility

water: 10%

shipped in

ambient

storage temp.

−20°C

InChI

1S/C22H40O12/c1-2-3-4-5-6-7-8-9-15(26)34-22(20(31)18(29)16(27)13(10-23)33-22)21(12-25)19(30)17(28)14(11-24)32-21/h13-14,16-20,23-25,27-31H,2-12H2,1H3/t13-,14-,16-,17-,18+,19+,20-,21+,22+/m1/s1

InChI key

KQXPBHIAPSOIPX-RAGLHBGGSA-N

General description

A non-ionic detergent useful for solubilization and isolation of pigment proteins of brown algae. Can be easily removed by dialysis.
A non-ionic detergent useful for solubilization and isolation of pigment proteins of brown algae. Can be easily removed by dialysis. Has a CMC of 2.5 mM.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 4 months at -20°C.

Other Notes

Nifant‘ev, N.E., et al. 1993. Carbohydr. Res. 250, 211.
Makimo, S., et al. 1983. Agric.Biol. Chem. 47, 319.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service