Merck
2633601
Supelco
Inlet-Liner, Splitless-Typ, gerades Design (ungepackt)
pkg of 1 ea
About This Item
UNSPSC-Code:
41115719
Verpackung
pkg of 1 ea
Hersteller/Markenname
Shimadzu 221-41444-01 (OEM)
L × AD × ID
95 mm × 5.0 mm × 3.4 mm
Kompatibilität
technique used for General purpose
Allgemeine Beschreibung
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Shimadzu (17A) [with SPL-17 Injector]
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)