Z744124
Lab-Sorb™ kit
with 25 small bags
Synonym(e):
spill control, spill granules
Alle Fotos(3)
About This Item
UNSPSC-Code:
47131905
Empfohlene Produkte
Verpackung
pack of 25 ea
Hersteller/Markenname
Diversified Biotech SORB-2000
Allgemeine Beschreibung
LabSorb kit with 25 small bags, converts liquid waste to a sold, for quick and easy disposal
Rechtliche Hinweise
Lab-Sorb is a trademark of Diversified Biotech
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)