Merck
Z743324
Breathe-EASY® petri dish strips, gas permeable polyurethane membrane
L × W 11.125 in. × 0.625 in.
Synonym(e):
breathable sealing membrane, cell culture, microplates, microwell plates, multi-well plates, multi-well sealing film, plate sealers, plate sealing film, sealing film, tissue culture
About This Item
UNSPSC-Code:
41121812
NACRES:
NB.35
Verpackung
pack of 100 ea
Hersteller/Markenname
Diversified Biotech BEMP-1000
L × B
11.125 in. × 0.625 in.
Allgemeine Beschreibung
Breathe-Easy® breathable strips for sealing petri and cell culture dishes. Allows the exchange of oxygen and carbon dioxide for consistant cellular growth.
Rechtliche Hinweise
Breathe-Easy is a registered trademark of Diversified Biotech
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Kunden haben sich ebenfalls angesehen
Slide 1 of 1