Merck
Z740410
SecurTainer II
tamper evident, volume 120 mL
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.22
Materialien
polyethylene cap
polypropylene container
Verpackung
pack of 300 ea
Hersteller/Markenname
Simport C576-120MA
Volumen
120 mL
Allgemeine Beschreibung
This model uses a different tamper evident concept when compared to the C575 Series. No locking tab is necessary when the tamper evident feature is used. Simply screw the cap on the container and the tamper evident sealing ring is automatically locked in place. When unscrewed, the ring is detached from the cap, showing clearly that the container was opened.
Also designed for collection, transport and storage of specimens, the polypropylene containers are safe to use even in adverse conditions. Containers are uniquely stackable, shatter resistant and are manufactured from virgin, polypropylene. These straight sided containers are available in many sizes from 20 to 120 ml. Non sterile. Containers and caps packaged separately in bags of 100.
Rechtliche Hinweise
SecurTainer is a trademark of Simport
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)