Z740391
The SpecTainer™ I, tamper evident
size 90 mL, non-sterile
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.22
Materialien
polyethylene cap
polypropylene container
Sterilität
non-sterile
Verpackung
pack of 400 ea
Hersteller/Markenname
Simport C566-90Y
Größe
90 mL
Verwandte Kategorien
Allgemeine Beschreibung
For applications needing the utmost security where sample integrity is of high importance. A great feature of the Spectainer I is that it incorporates a unique tamper evident screw cap ensuring peace of mind during transport or storage situations where someone might have tampered with the specimen.
Both containers and caps are manufactured without the use of plasticisers or mold release agents. All material used is free from latex. Containers are 95kpa compliant. Available in three sizes: 60, 90 and 120 ml. The 60 ml size however is not graduated. The sterile container protects its sterility with a tamper evident seal.
Both containers and caps are manufactured without the use of plasticisers or mold release agents. All material used is free from latex. Containers are 95kpa compliant. Available in three sizes: 60, 90 and 120 ml. The 60 ml size however is not graduated. The sterile container protects its sterility with a tamper evident seal.
Rechtliche Hinweise
SpecTainer is a trademark of Simport
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)