Z740252
TubePopper® for one-handed opening of microtubes
pkg of 1 ea
Alle Fotos(2)
About This Item
UNSPSC-Code:
41121702
NACRES:
NB.01
Empfohlene Produkte
Verpackung
pkg of 1 ea
Hersteller/Markenname
Diversified Biotech TPOP-1000
Allgemeine Beschreibung
Opening and closing points are dimpled to fit domed tubes. Manufactured in clear acrylic.
Rechtliche Hinweise
T3P is a registered trademark of Archimica GmbH
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)