Merck
Z709603
Sartorius mLINE® mechanical pipettor
8 channel, volume 5-100 μL
Synonym(e):
Biohit pipettes, pipettor
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Leistungsmerkmale
8-channel
electronic: no
Verpackung
pack of 1 ea
Hersteller/Markenname
Sartorius 725130
Messbereich Volumen
5-100 μL
Volumen
100 μL
Allgemeine Beschreibung
Sartorius′s new mechanical pipettor family - the mLINE - offers a unique combination of novel features for mechanical pipetting. Excellent performance, full autoclavability, smooth ergonomics and perfect safety. The mLINE pipettors cover the full volume range of 0.1 to 5000.
  • Extremely light pipetting and tip ejection
  • Unique safety features
  • Stepless volume adjustment and patented volume locking system
  • Thermal insulation of internal components to improve accuracy
  • Full autoclavability without disassembling
  • Easy cleaning, maintenance and calibration
  • ID tag and color-coded caps
  • Safe-Cone Filters and unique ejection
  • Opti-load tip mounting mechanism for even tip sealing and light tip ejection in multichannel pipettors
Rechtliche Hinweise
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)