Merck
Z617954
Greiner culture tubes
14 mL round bottom culture tube, polypropylene, sterile, w/ graduation, w/ snap (vent) cap
Synonym(e):
culture tubes, culture vials, plastic culture tubes, Greiner culture tubes
About This Item
UNSPSC-Code:
41121706
NACRES:
NB.22
Materialien
colorless tube
conical cap (Push fit)
low-density polyethylene cap
polypropylene tube
round bottom
straight neck
Sterilität
sterile
Leistungsmerkmale
RCF 4,800 × g
neck thread push fit
Verpackung
pack of 800 ea (individually wrapped)
Hersteller/Markenname
Greiner 187261
Kapazität
14 mL
Durchm. × L
18 mm × 95 mm
Allgemeine Beschreibung
Greiner culture tubes feature a two-position snap vent stopper and graduations with a writing area.
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)