TMO4112-1000
Nalgene® sterile single-use Erlenmeyer flask with solid closure
PETG, bottle volume 1000 mL, case of 6 ea
Synonym(e):
Nalgene Erlenmeyer flask, single use plastic flask
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.13
Empfohlene Produkte
Materialien
PETG
Sterilität
sterile
Verpackung
case of 6 ea
Hersteller/Markenname
Nalgene 4112-1000
Flaschenvolumen
1000 mL
Allgemeine Beschreibung
Save preparation time and avoid contamination risk by using Thermo Scientific Nalgene Single-Use PETG Erlenmeyer Flasks with Baffled Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.
Sonstige Hinweise
Browse all Erlenmeyer flasks
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)