Merck
TMO342002-9025
Nalgene® sterile diagnostic bottle
high-density polyethylene, bottle capacity 8 mL, natural, case of 980 ea
Synonym(e):
small volume plastic bottle, test kit bottle
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
high-density polyethylene
natural
Sterilität
sterile
Verpackung
case of 980 ea
Hersteller/Markenname
Nalgene 342002-9025
Flaschenkapazität
8 mL
Allgemeine Beschreibung
Perform many small volume packing applications with Thermo Scientific Nalgene HDPE Diagnostic Bottles with Closure. Bottles can be used in test kits and are packed in shrink-wrapped SBS trays for easy handling. Trays can be used as an aid for machine fill-line loading. They come assembled with closures without a liner that can wrinkle.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
TMO342002-90254061841584476