Merck
TMO312751-9125
Nalgene® Dropper Bottle without tip or cap
white low-density polyethylene, volume 4 mL, case of 2000 ea
Synonym(e):
LDPE dropper bottle, dispensing bottle
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
white low-density polyethylene
Verpackung
case of 2000 ea
Hersteller/Markenname
Nalgene 312751-9125
Kapazität
4 mL
Volumen
4 mL
Allgemeine Beschreibung
Repeatedly carry out reliable routine dispensation with Thermo Scientific Nalgene LDPE Dropper Bottles, made of low-density polyethylene. These bottles are to be used with Nalgene controlled dropper tips and closures.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)