Merck
TMO2118-0001
Jars Nalgene® style 2118
clear polypropylene copolymer, 30 mL capacity, case of 72 ea
Synonym(e):
laboratory jars, nalgene jars
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
clear polypropylene copolymer
polypropylene bottle
Beschreibung
Autoclavable with closure completely unscrewed
Verpackung
case of 72 ea
Hersteller/Markenname
Nalgene 2118-0001
Kapazität
30 mL
30 mL capacity
Allgemeine Beschreibung
Translucent with excellent chemical resistance.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries