Merck
TMO2116-0500
Nalgene® straight-sided wide-mouth jar
polycarbonate, clear, case of 16 ea, jar capacity 500 mL
Synonym(e):
clear plastic container, screw top container
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
clear
polycarbonate
straight
wide-mouth
Verpackung
case of 16 ea
Hersteller/Markenname
Nalgene 2116-0500
Becher-Fassungsvermögen
500 mL
Allgemeine Beschreibung
Collect and store pastes, solids, specimens and lab supplies with window-clear Thermo Scientific Nalgene Straight-side Wide-Mouth Jars. A wide temperature range (-135°C to +135°C) makes these jars an excellent choice for cold room and refrigerated use all the way to the autoclave. These high impact-resistant jars are extremely versatile. Not liquid leakproof.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)