TMO150067
Nunc® Thermanox™ coverslip
Thermanox™ plastic, L × W 24 mm × 30 mm, sterile, case of 500 ea
Synonym(e):
cell culture coverslip, cell culture treated coverslip
Alle Fotos(1)
About This Item
UNSPSC-Code:
41161502
NACRES:
NB.13
Empfohlene Produkte
Sterilität
sterile
Verpackung
case of 500 ea
Hersteller/Markenname
Nunc 150067
L × B
24 mm × 30 mm
Verwandte Kategorien
Allgemeine Beschreibung
Experience the best features of the Thermo Scientific storage tube portfolio: universal cap, universal latch rack, and free-standing tube. This versatile 2.0mL internally-threaded tube is ideal both for a large storage system like a biobank, and in smaller scale for traditional cryostorage. **Now available in a barcoded rack to allow tracking of the rack in addition to individual tubes. Sample identity is ensured due to a permanently attached 2D barcode on each tube and permanent laser etching of the rack. All tubes and racks are supplied free from DNA, RNase, DNase, and endotoxins.
Rechtliche Hinweise
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Thermanox is a trademark of Thermo Fisher Scientific, Inc
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
TMO150067 |