Merck
TMO140360
Nunc® EasyFill Cell Factory System
4 layer, polystyrene, surface area 2528 cm2, sterile, vented, case of 4 ea
Synonym(e):
closed cell system layered storage rack, closed cell system rack
About This Item
UNSPSC-Code:
41161502
NACRES:
NB.13
Materialien
polystyrene
Sterilität
sterile
Leistungsmerkmale
vented
Verpackung
case of 4 ea
Hersteller/Markenname
Nunc 140360
Oberflächenbereich
2528 cm2
Bindungsart
Nunclon Delta surface
Allgemeine Beschreibung
The Nunc® EasyFill Cell Factory systems are a space saving, out-of-the-box, multilayer solution and are as easy to use as a T-flask. Scale up your research and commercial-scale applications with confidence. These systems come with a cerrtified Nunclon® Delta cell culture surface to enable consistent performance from lot to lot and format to format. The 10-layer, single-use system holds the equivalent of 36 T175 flasks.
Leistungsmerkmale und Vorteile
  • "Out of the box" system with no accessories required.
  • Large, vented screw closure for easy handling
  • Versatile port design allows for pouring and aseptic filling.
  • Fill and empty five times faster compared to T175 flasks.
  • Enhances productivity by enabling faster operations.
  • Fast start-up with a simple pour of media and cells into the large opening.
  • Versatile port facilitates pouring and aseptic filling techniques.
Rechtliche Hinweise
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
TMO1403604061841580416