OH30073983
Ohaus® Pen pH Meter
Model ST20S
Alle Fotos(1)
About This Item
UNSPSC-Code:
41115603
Empfohlene Produkte
Materialien
epoxy shaft
Verpackung
pack of 1 ea
Hersteller/Markenname
Ohaus® 30073983
Messbereich Salzgehalt
0.0-80.0 ppt
Temperaturmessbereich
0.0-99.0 °C
Akkuleistung
4 × 1.5 V, 200 hr life
Allgemeine Beschreibung
With durable IP67 waterproof ABS housing, protective sensor cap, and an automatic shutdown feature that preserves battery life, OHAUS pen meters can endure consistent use in rough, wet environments.
Anwendung
pH, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), salinity, total dissolved solids (TDS), temperature measurement
Leistungsmerkmale und Vorteile
Easily replaceable electrodes, automatic shut off feature
Rechtliche Hinweise
Ohaus is a registered trademark of Ohaus Corp.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)