Merck
MTCP1001QPS
MTC Bio Pure.Amp qPCR Sealing Film
to fit standard qPCR plates, pressure adhesive
Synonym(e):
qPCR sealing membrane, sealing membrane, snap closure tube
About This Item
UNSPSC-Code:
41122410
NACRES:
NB.15
Verpackung
bag of 10 × 10 ea
Hersteller/Markenname
MTC Bio P1001-QPS
Verwandte Kategorien
Allgemeine Beschreibung
Sealing film, qPCR Optical (pressure-sensitive adhesive) ABI Type, 10 bags of 10 seals, 100/pk
Leistungsmerkmale und Vorteile
  • RNase, DNase, DNA, and PCR inhibitor free
  • Evaporation rates ≤ 5%
  • Inner packs of 10 membranes
  • -40° to 120°C
Rechtliche Hinweise
MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)