Merck
MTCA8813
MTC Bio Blu-Lite UHC Autoradiography Flim
W × L 5 in. × 7 in.
Synonym(e):
blotting film, western imaging film
About This Item
UNSPSC-Code:
41105341
NACRES:
NB.15
Verpackung
pack of 100 ea
Hersteller/Markenname
MTC Bio A8813
B × L
5 in. × 7 in.
Allgemeine Beschreibung
Blu-Lite UHC Autoradiography film, 5x7in, 100 sheets/box
Leistungsmerkmale und Vorteile
  • Medium/dark blue base
  • Ideal for chemiluminescence
  • Ultra-high contrast
  • Ideal for 14C, 32P, 33P, 125I, and 35S
Rechtliche Hinweise
Blu-Lite is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)