Merck
HS4283P
Microtube Storage Box
to hold, 50 tube(s), white expanded polystyrene rack
About This Item
UNSPSC-Code:
41122800
Materialien
white expanded polystyrene rack
Leistungsmerkmale
to hold 50 tube(s)
Verpackung
pack of 10 ea
Hersteller/Markenname
Heathrow Scientific HS4283P
L × B × H
210 mm × 110 mm × 71 mm
Allgemeine Beschreibung
The Foam Tube Rack provides high storage with minimal footprint.
Leistungsmerkmale und Vorteile
  • Two-piece racks are constructed of expanded polystyrene (EPS) foam
  • Lightweight microtube racks are ideal for long-term studies or tube storage
  • Extra spacing between wells allows for easy gripping with fingertips
  • Racks stack securely with nesting features on lid and base
  • Store down to temperatures of -80°C. Must use tubes capable of being stored at the required temperature of the user
  • Not recommended for autoclaving
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)