GE11002688
GE Ettan™ IPGphor™ manifold light kit
complete
Alle Fotos(1)
About This Item
UNSPSC-Code:
41105300
NACRES:
NB.22
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
Cytiva 11-0026-88
Cytiva Article No. 11-0026-88
Leistungsmerkmale und Vorteile
- For Use with Ettan™ IPGphor™ 3 isoelectric focusing unit
- Includes: 1 x Cleaning brush, 2 x Electrode assembly, 1 x Spirit level, 20 x Sample cup, 1 x Sample cup insertion tool, 60 x Paper electrode wicks, 60 x Paper bridge pads, 1 x Manifold, 1 x Forceps,1 x 60 ml cleaning, solution and User Manual
Rechtliche Hinweise
Ettan is a trademark of Cytiva
IPGphor is a trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)