GBL664113
Grace Bio-Labs FastWells™ reagent barriers
1- 20 mm diam. x 1.0 mm depth
Synonym(e):
flexible silicone slide gasket, reagent chamber
Alle Fotos(2)
About This Item
UNSPSC-Code:
41122600
NACRES:
NB.22
Empfohlene Produkte
Leistungsmerkmale
wells 1
Verpackung
pack of 50 ea
Hersteller/Markenname
Grace Bio-Labs 664113
Außenmaß
25 mm × 25 mm
Allgemeine Beschreibung
FastWells reagent barriers are sticky, flexible silicone gaskets that form hydrophobic barriers around specimens without messy adhesives or special slides. Gaskets may be stacked to increase depth and volume.
FastWells provide rapid isolation of cells and tissues on slides or coverslips for antibody incubations. The barriers remain sealed during agitated washing steps, preventing reagents from spilling when shaken or rocked. FastWells peel off quickly and cleanly, leaving no residue to interfere with specimen cover-slipping. They can be washed, autoclaved and reused. The barriers can also be sealed to form incubation chambers using flexible HybriSlip covers.
FastWells provide rapid isolation of cells and tissues on slides or coverslips for antibody incubations. The barriers remain sealed during agitated washing steps, preventing reagents from spilling when shaken or rocked. FastWells peel off quickly and cleanly, leaving no residue to interfere with specimen cover-slipping. They can be washed, autoclaved and reused. The barriers can also be sealed to form incubation chambers using flexible HybriSlip covers.
Rechtliche Hinweise
FastWells is a trademark of Grace Bio-Labs, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
GBL664113-50EA | 4061832399935 |