Merck
EP5920751000
Adapter for Eppendorf® S-4x Universal Large Rotor
capacity, 24 x 14 mL tubes, pack of 2
About This Item
UNSPSC-Code:
41103911
NACRES:
NB.82
Leistungsmerkmale
capacity 24 x 14 mL tubes
Holds 14 mL tubes
Verpackung
pack of 2
Hersteller/Markenname
Eppendorf® 5920751000 (Global)
Methode(n)
centrifugation: suitable
Kompatibilität
for use with Universal-Large bucket
Allgemeine Beschreibung
Adapters for various sizes of tubes and bottles for use with Eppendorf S-4x Universal large rotor.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)