Merck
EP5810757002
Eppendorf® Adapter for 250 mL Rectangular Bucket
for use with 4 x 18-30 mL tubes, set of 2 pieces
About This Item
UNSPSC-Code:
41103911
NACRES:
NB.82
Leistungsmerkmale
for use with 4 x 18-30 mL tubes
Verpackung
set of 2 pieces
Hersteller/Markenname
Eppendorf® 5810757002
Methode(n)
centrifugation: suitable
Kompatibilität
for use with 250 mL rectangular buckets
Allgemeine Beschreibung
Tube adapter for Eppendorf 250 mL rectangular bucket.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG