EP4926000026
Eppendorf® Reference® 2 Variable Volume Pipettor
12-channel, incl. epT.I.P.S.® Box, volume 0.5-10 μL, gray
Alle Fotos(4)
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Empfohlene Produkte
Materialien
gray
Leistungsmerkmale
12-channel
mechanical
Verpackung
pack of 1 ea
Hersteller/Markenname
Eppendorf® 4922000021 (Global)
Volumen
0.5-10 μL
Verwandte Kategorien
Allgemeine Beschreibung
The Reference 2 is the successor to the highly successful Reference pipette, and enhances the range′s already high standards with a new design, reduced weight and operating forces and multi-channel versions. As the premium product in Eppendorf′s pipette portfolio, the Reference 2 provides the most accurate results possible alongside robust, reliable handling and optimum user safety.
These levels of security make the Reference 2 ideal for use with precious liquids, or in any application requiring extremely high levels of accuracy. When there can be no compromise on quality and reproducability, Eppendorf′s new Reference 2 is an ideal instrument for these liquid handling applications.
These levels of security make the Reference 2 ideal for use with precious liquids, or in any application requiring extremely high levels of accuracy. When there can be no compromise on quality and reproducability, Eppendorf′s new Reference 2 is an ideal instrument for these liquid handling applications.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
EP4926000026-1EA | 4061835301805 |