Merck
EP2231302005
Eppendorf® Reference 2 Variable Volume Pipettor
new
no. of channels, Single Channel, pack of 4, volume 0.5—10 μL, volume 2—20 μL, volume 20—200 μL, volume 100—1,000 μL, includes tips
Synonym(e):
Racks
About This Item
UNSPSC-Code:
41121500
Leistungsmerkmale
adjustable
manual
no. of channels Single Channel
Verpackung
pack of 4
Hersteller/Markenname
Eppendorf® 2231302005
Volumen
0.5—10 μL
100—1,000 μL
20—200 μL
2—20 μL
Verwandte Kategorien
Allgemeine Beschreibung
Eppendorf® Reference 2, 4-pack Option 3, 1-channel, variable, includes 4 adjustable-volume pipettes (0.5–10 µL, 2–20 µL/yellow, 20–200 µL, 100–1,000 µL), 1 full box of Eppendorf pipette tips for each pipette volume (excludes 5 mL and 10 mL tips) , 0.1 µL – 10 mL.
Leistungsmerkmale und Vorteile
  • Mechanical air-cushion pipette with outstanding precision and accuracy - operated with just one button for volume selection, pipetting and tip ejection
  • With its innovative one-button operation, the Reference 2 allows fast and ergonomic handling with reduced operating efforts and active aerosol reduction.
  • A clear haptic feedback helps to avoid accidental tip ejection 4-digit display with magnifying window for quick and secure volume setting and identification (clearly visible from every angle), incl. volume lock
  • Spring-loaded tip cones improve user-to-user reproducibility and ergonomics by lowering the tip attachment force
  • The unique smooth surface and full autoclavability guarantee efficient decontamination - making it the ideal companion when working under sterile conditions
  • A temporary adjustment allows for an easy adjustment of your Reference 2 for the most accurate pipetting of different liquids or in changing external conditions without the need for a full calibration
  • Available as single-channel pipette in fixed or variable volume as well as 8- and 12-channel pipette
  • The Reference 2 multi-channel variants offer high flexibility incl. a spring-loaded tip cone which can be switched on/off optionally
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)