CLS7007525
Pyrex® VISTA™ graduated cylinder, to contain, economy
volume 25 mL
Synonym(e):
lab equipment accessory
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121805
NACRES:
NB.43
Empfohlene Produkte
Hersteller/Markenname
Corning 70075-25
Volumen
25 mL
Allgemeine Beschreibung
These Pyrex VISTA cylinders are calibrated "to contain" (TC). They are supplied in two parts - a strong, accurate glass graduate and a detachable, sturdy plastic base which absorbs impacts and reduces breakage. Tolerances are +/- 5% of total at any point. By removing the base, the graduate can be conveniently stacked. Bumper guards are supplied with 25 mL through 100 mL inclusive. The 10 mL has a funnel top for careful pouring.
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)