CLS6796400D
Corning® Digital Hotplates
AC/DC input 230 V AC (Supplied with both 3-pin UK plug and 2-pin Euro plug), plate L × W 5 in. × 7 in., CE compliant
Synonym(e):
stirrers
Alle Fotos(2)
About This Item
UNSPSC-Code:
41102406
NACRES:
NB.72
Empfohlene Produkte
Leistungsmerkmale
CE compliant
Verpackung
case of 1
Hersteller/Markenname
Corning 6796-400D
Verfügbarkeit
not available in USA and Canada
Parameter
5-550 °C temp. range
Platte L × B
5 in. × 7 in.
Allgemeine Beschreibung
- Digital LED temperature display is adjustable in 5 °C increments and blinks until set temperature is reached
- Microprocessor maintains consistent and repeatable temperature settings.
- Bright LED HOT TOP icon lights up when top plate temperature is over 60 °C, even when heat control is turned off.
- Separate temperature sensor provides power cut off if unit overheats
- Extremely durable and heat resistant Pyroceram glass-ceramic top
- Small footprint and low profile
- Meets UL and cUL standards
- 2 year warranty
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)