CLS431845
Corning® centrifuge bottle
with standard screw cap, capacity 500 mL, polypropylene
Synonym(e):
centrifugation bottles, digestion bottles, shaking bottles
About This Item
Empfohlene Produkte
Materialien
flat bottom tube
polypropylene
round bottle
translucent bottle
wide-mouth bottle
Sterilität
non-sterile
Leistungsmerkmale
autoclavable
cap
Verpackung
pkg of 24 ea
Hersteller/Markenname
Corning 431845
Parameter
13700 × g max. RCF
Kapazität
500 mL
Verwandte Kategorien
Leistungsmerkmale und Vorteile
- Corning® centrifuge bottles have a specially designed cap for improved grip
- Includes clear graduations and textured marking spot.
- Bottles are designed with extra-thick walls for high durability
- Wide mouth for easier liquid filling
- Compatible with most centrifuges and do not require special buckets
- Unique design aid researchers to reduce the number of centrifugation cycles and enhance productivity
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
CLS431845-24EA | 4061833885673 |