CLS10042
Corning® HYPERStack® disposable tubing set
for use with glass bottle, sterile
Synonym(e):
lab equipment accessory
Alle Fotos(1)
About This Item
UNSPSC-Code:
41105108
NACRES:
NA.75
Empfohlene Produkte
Beschreibung
18 in. C-Flex® tubing
Sterilität
sterile
Verpackung
pkg of 2 ea
Hersteller/Markenname
Corning 10042
Rechtliche Hinweise
C-Flex is a registered trademark of Saint-Gobain Performance Plastics Corp.
Corning is a registered trademark of Corning, Inc.
HYPERStack is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)